Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecahydrodibenzofuran (mixture of isomers) ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

D5232-25G D5232-5G
TCD5232-25GEA 575 USD InStock
TCD5232-25G TCD5232-5G
Dodecahydrodibenzofuran (mixture of isomers) ≥98.0% (by GC)
Dodecahydrodibenzofuran (mixture of isomers)


Formula: C₁₂H₂₀O
Boiling Pt: 259°C
CAS Number: 13054-98-3
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty