Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylammonium tetrakis(trifluoromethyl)cuprate(III) ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd
Warning

T3374-1G T3374-200MG
TCT3374-1GEA 310 USD InStock
TCT3374-1G TCT3374-200MG
Tetrabutylammonium tetrakis(trifluoromethyl)cuprate(III) ≥98.0% (by titrimetric analysis)
Tetrabutylammonium tetrakis(trifluoromethyl)cuprate(III)


Formula: C₂₀H₃₆CuF₁₂N
MW: 582.04 g/mol
Melting Pt: 142°C
CAS Number: 152376-94-8
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty