Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-tetrazole ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

M2451-1G M2451-5G
TCM2451-1GEA 97 USD InStock
TCM2451-1G TCM2451-5G
1-Methyl-1H-tetrazole ≥98.0% (by GC)
1-Methyl-1H-tetrazole


Formula: C₂H₄N₄
Boiling Pt: 81°C
MDL Number: MFCD00129972
CAS Number: 16681-77-9
UN: 1325
ADR: 4.1,III
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty