Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyldimethylsilanol ≥95.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

B5499-25ML B5499-5ML
TCB5499-25MLEA 273 USD InStock
TCB5499-25ML TCB5499-5ML
tert-Butyldimethylsilanol ≥95.0% (by GC)
tert-Butyldimethylsilanol


Formula: C₆H₁₆OSi
MW: 132.28 g/mol
Boiling Pt: 139°C
Flash Pt: 45°C
MDL Number: MFCD00010673
CAS Number: 18173-64-3
UN: 1987
ADR: 3 III
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty