Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2-Acetoxypropionic acid ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

A3084-1G A3084-5G
76218-790EA 67 USD InStock
76218-790 76218-792
(R)-(+)-2-Acetoxypropionic acid ≥98.0% (by GC, titration analysis)
(R)-(+)-2-Acetoxypropionic acid


Formula: C₅H₈O₄
Boiling Pt: 82°C
Storage Temperature: Refrigerator
MDL Number: MFCD16036178
CAS Number: 18668-00-3
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 97.0 ee%
Purity (Neutralization titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty