Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,4-dimethylphenyl)amine ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

B5148-1G B5148-200MG
TCB5148-1GEA 310 USD InStock
TCB5148-1G TCB5148-200MG
Bis(2,4-dimethylphenyl)amine ≥98.0% (by GC)
Bis(2,4-dimethylphenyl)amine


Formula: C₁₆H₁₉N
Boiling Pt: 310°C
MDL Number: MFCD16619146
CAS Number: 19616-28-5
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty