Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-p-Toluenesulfinamide 98,0 HPLC_ASSAY_METHOD

Supplier: Decon Laboratories Ltd

T2814-1G T2814-200MG
76220-528EA 285 USD InStock
76220-528 76220-530
(R)-(-)-p-Toluenesulfinamide 98,0 HPLC_ASSAY_METHOD
(R)-(-)-p-Toluenesulfinamide


Formula: C₇H₉NOS
MW: 155.22 g/mol
MDL Number: MFCD06858374
CAS Number: 247089-85-6
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 116.0 to 120.0 deg-C
Specific rotation [a]20/D -81.0 to -91.0 deg (C=1, CHCL3)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty