Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd
Warning

D5245-250MG D5245-50MG
76219-870EA 444 USD InStock
76219-870 76219-872
2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine ≥98.0% (by HPLC)
2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine


Formula: C₇H₁₁N₅O₂
Storage Temperature: Freezer
MDL Number: MFCD18379166
CAS Number: 252944-01-7
Order Now
Specification Test Results
Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty