Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Titanyl Phthalocyanine, purified by sublimation

Supplier: Decon Laboratories Ltd
Warning

T2272-1G T2272-200MG
TCT2272-1GEA 244 USD InStock
TCT2272-1G TCT2272-200MG
Titanyl Phthalocyanine, purified by sublimation
Titanyl Phthalocyanine


Formula: C₃₂H₁₆N₈OTi
MW: 576.40 g/mol
Melting Pt: 395°C
MDL Number: MFCD00145414
CAS Number: 26201-32-1
EINECS: 419-970-5
Order Now
Specification Test Results
Appearance Dark red to Dark purple to Dark blue powder to crystal
Elemental analysis (Nitrogen) 18.50 to 20.00 %
Lambda max. 692.0 to 694.0 nm (Pyridine)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty