Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alibendol ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

A2965-1G A2965-5G
TCA2965-1GEA 21 USD InStock
TCA2965-1G TCA2965-5G
Alibendol ≥98.0% (by GC)
Alibendol


Formula: C₁₃H₁₇NO₄
MDL Number: MFCD00865165
CAS Number: 26750-81-2
EINECS: 247-960-9
Merck Index: 14,00244
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty