Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisopropylcyanamide 98,0 GC_ASSAY_METHOD

Supplier: Decon Laboratories Ltd
Warning

D4952-1ML D4952-5ML
TCD4952-1MLEA 317 USD InStock
TCD4952-1ML TCD4952-5ML
Diisopropylcyanamide 98,0 GC_ASSAY_METHOD
Diisopropylcyanamide


Formula: C₇H₁₄N₂
MW: 126.20 g/mol
Boiling Pt: 94°C
Melting Pt: -27°C
MDL Number: MFCD00008865
CAS Number: 3085-76-5
EINECS: 221-401-9
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty