Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fenofibric acid ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

F1011-1G F1011-5G
TCF1011-1GEA 29 USD InStock
TCF1011-1G TCF1011-5G
Fenofibric acid ≥98.0% (by GC, titration analysis)
Fenofibric acid


Formula: C₁₇H₁₅ClO₄
MDL Number: MFCD00792461
CAS Number: 42017-89-0
EINECS: 255-626-9
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 179.0 to 183.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty