Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetoxypropionic acid ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd
Warning

A2931-1G A2931-5G
TCA2931-1GEA 48.1 USD InStock
TCA2931-1G TCA2931-5G
2-Acetoxypropionic acid ≥98.0% (by titrimetric analysis)
2-Acetoxypropionic acid


Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 127°C
Storage Temperature: Refrigerator
MDL Number: MFCD00674491
CAS Number: 535-17-1
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty