Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethynyluracil ≥97.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

E1096-1G E1096-200MG
TCE1096-1GEA 705 USD InStock
TCE1096-1G TCE1096-200MG
5-Ethynyluracil ≥97.0% (by HPLC, titration analysis)
5-Ethynyluracil


Formula: C₆H₄N₂O₂
Melting Pt: 320°C
MDL Number: MFCD00871973
CAS Number: 59989-18-3
Merck Index: 14,03584
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty