Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pamabrom ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

P2359-1G P2359-5G
TCP2359-1GEA 17 USD InStock
TCP2359-1G TCP2359-5G
Pamabrom ≥98.0% (by HPLC, titration analysis)
Pamabrom


Formula: C₁₁H₁₈BrN₅O₃
Melting Pt: 295°C
MDL Number: MFCD00867328
CAS Number: 606-04-2
EINECS: 210-103-4
Merck Index: 14,07000
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty