Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',5-Dichloro-2-hydroxybenzophenone ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd
Warning

D4703-1G D4703-5G
76219-058EA 47 USD InStock
76219-058 76219-060
2',5-Dichloro-2-hydroxybenzophenone ≥98.0% (by GC)
2',5-Dichloro-2-hydroxybenzophenone


Formula: C₁₃H₈Cl₂O₂
MW: 267.11 g/mol
MDL Number: MFCD02682994
CAS Number: 61785-35-1
EINECS: 262-965-6
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty