Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lobenzarit disodium salt ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

L0278-100MG L0278-25MG
TCL0278-100MGEA 338 USD InStock
TCL0278-100MG TCL0278-25MG
Lobenzarit disodium salt ≥98.0% (by HPLC, titration analysis)
Lobenzarit disodium salt


Formula: C₁₄H₈ClNNa₂O₄
Melting Pt: 388°C
MDL Number: MFCD00941422
CAS Number: 64808-48-6
Merck Index: 14,05552
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty