Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cycloastragenol

Supplier: Decon Laboratories Ltd

C3469-100MG C3469-25MG
TCC3469-100MGEA 90 USD InStock
TCC3469-100MG TCC3469-25MG
Cycloastragenol
Cycloastragenol


Formula: C₃₀H₅₀O₅
Melting Pt: 241°C
Storage Temperature: Refrigerator
MDL Number: MFCD24849201
CAS Number: 78574-94-4
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 241.0 to 245.0 deg-C
Specific rotation [a]20/D +50.0 to +55.0 deg (C=1, methanol)

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty