Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

F1083-1G F1083-5G
76220-082EA 95 USD InStock
76220-082 76220-084
1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid ≥98.0% (by HPLC, titration analysis)
1-[(4-Fluorophenyl)carbamoyl]cyclopropanecarboxylic acid


Formula: C₁₁H₁₀FNO₃
MW: 223.20 g/mol
MDL Number: MFCD11226313
CAS Number: 849217-48-7
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty