Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Roxatidine Acetate hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

R0178-1G R0178-200MG
TCR0178-1GEA 116 USD InStock
TCR0178-1G TCR0178-200MG
Roxatidine Acetate hydrochloride ≥98.0% (by HPLC, titration analysis)
Roxatidine Acetate hydrochloride


Formula: C₁₉H₂₈N₂O₄·HCl
MW: 384.90 g/mol
MDL Number: MFCD00941429
CAS Number: 93793-83-0
Merck Index: 14,08274
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty