Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Diaminotoluene ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Danger

D4628-25G D4628-5G
TCD4628-25GEA 259 USD InStock
TCD4628-25G TCD4628-5G
2,5-Diaminotoluene ≥98.0% (by GC, titration analysis)
2,5-Diaminotoluene


Formula: C₇H₁₀N₂
Boiling Pt: 168°C
MDL Number: MFCD00035779
CAS Number: 95-70-5
EINECS: 202-442-1
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty