Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EN450 98%

Supplier: AMBEED, INC
Warning

A1449227-10MG A1449227-5MG
77622-154EA 95.79 USD InStock
77622-154 77622-156
EN450 98%
EN450
Precaución: for Research Use Only


Formula: C₁₁H₁₃ClN₂O₃S
MW: 288.75 g/mol
Storage Temperature: Refrigerator
CAS Number: 793719-01-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty