Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AMPC 98%

Supplier: AMBEED, INC
Warning

A1550222-10MG A1550222-5MG
77625-396EA 70.04 USD InStock
77625-396 77625-398
AMPC 98%
AMPC
Precaución: for Research Use Only


Formula: C₂₅H₁₆FN₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD34474080
CAS Number: 2254434-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty