Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(−)-Caryophyllene oxide 95%

Supplier: AMBEED, INC
Warning

A159928-250MG A159928-5G A159928-1G
77625-520EA 10.3 USD InStock
77625-520 77625-522 77625-518
(−)-Caryophyllene oxide 95%
(−)-Caryophyllene oxide
Precaución: for Research Use Only


Formula: C₁₅H₂₄O
Storage Temperature: Refrigerator
MDL Number: MFCD00134216
CAS Number: 1139-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty