Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lovastatin hydroxy acid sodium 99%

Supplier: AMBEED, INC
Warning

A398908-10MG A398908-5MG
77668-830EA 84 USD InStock
77668-830 77668-832
Lovastatin hydroxy acid sodium 99%
Lovastatin hydroxy acid sodium
Precaución: for Research Use Only


Formula: C₂₄H₃₇O₆·Na
MW: 444.54 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00870475
CAS Number: 75225-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty