Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N6-Benzoyl-2',3'-isopropylidene adenosine 97%

Supplier: AMBEED, INC
Warning

A928404-25G A928404-5G
77623-156EA 175.1 USD InStock
77623-156 77623-158
N6-Benzoyl-2',3'-isopropylidene adenosine 97%
N6-Benzoyl-2',3'-isopropylidene adenosine
Precaución: for Research Use Only


Formula: C₂₀H₂₁N₅O₅
Storage Temperature: Refrigerator
MDL Number: MFCD15145205
CAS Number: 39947-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty