Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl(2,4,6-trifluorophenyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

118247-5G 118247-25G 118247-1G 118247-500MG
77631-100EA 1484.23 USD InStock
77631-100 77635-412 77640-422 77648-404
Phenyl(2,4,6-trifluorophenyl)methanol ≥97%
Phenyl(2,4,6-trifluorophenyl)methanol
Precaución: For Research Use Only


Formula: 238.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13243433
CAS Number: 1307117-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty