Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Formyl-1-naphthaleneboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

11947-5G 11947-25G 11947-1G 11947-500MG
77631-512EA 614 USD InStock
77631-512 77657-270 77640-458 77644-436
4-Formyl-1-naphthaleneboronic acid ≥95%
4-Formyl-1-naphthaleneboronic acid
Precaución: For Research Use Only


Formula: C₁₁H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD01632205
CAS Number: 332398-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty