Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluoronicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

13548-5G 13548-500MG 13548-25G 13548-1G
77631-584EA 727 USD InStock
77631-584 77652-340 77653-466 77640-580
2-Bromo-5-fluoronicotinic acid ≥97%
2-Bromo-5-fluoronicotinic acid
Precaución: For Research Use Only


Formula: C₆H₃BrFNO₂
MW: 219.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870208
CAS Number: 38186-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty