Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Methylbenzo[b]thiophen-2-yl)boronic acid ≥97%

Supplier: AOB CHEM USA

13581-5G 13581-1G 13581-25G 13581-500MG
77631-586EA 1218 USD InStock
77631-586 77636-568 77653-472 77648-580
(3-Methylbenzo[b]thiophen-2-yl)boronic acid ≥97%
(3-Methylbenzo[b]thiophen-2-yl)boronic acid
Precaución: For Research Use Only


Formula: C₉H₉BO₂S
MDL Number: MFCD08706273
CAS Number: 912331-27-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty