Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[b]thiophen-5-ol ≥97%

Supplier: AOB CHEM USA
Warning

171191-5G 171191-25G 171191-1G 171191-500MG
77634-176EA 374 USD InStock
77634-176 77657-710 77640-920 77648-950
Benzo[b]thiophen-5-ol ≥97%
Benzo[b]thiophen-5-ol
Precaución: For Research Use Only


Formula: C₈H₆OS
Storage Temperature: Ambient
MDL Number: MFCD16876937
CAS Number: 19301-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty