Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-dichloro-6-methoxybenzaldehyde ≥95%

Supplier: AOB CHEM USA

19922-5G 19922-1G 19922-25G 19922-500MG
77631-954EA 936 USD InStock
77631-954 77637-134 77657-908 77649-192
2,3-dichloro-6-methoxybenzaldehyde ≥95%
2,3-dichloro-6-methoxybenzaldehyde
Precaución: For Research Use Only


Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
MDL Number: MFCD20485513
CAS Number: 936249-28-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty