Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-bromo-3-methoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

229105-5G 229105-1G 229105-25G 229105-500MG
77634-424EA 342 USD InStock
77634-424 77637-308 77658-086 77645-290
tert-Butyl-4-bromo-3-methoxybenzoate ≥97%
tert-Butyl 4-bromo-3-methoxybenzoate
Precaución: For Research Use Only


Formula: C₁₂H₁₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD08436063
CAS Number: 247186-51-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty