Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-(1-methylethoxy)pyridine ≥95%

Supplier: AOB CHEM USA
Warning

229678-5G 229678-1G 229678-25G 229678-500MG
77632-088EA 1151 USD InStock
77632-088 77637-312 77654-258 77649-346
2-Fluoro-6-(1-methylethoxy)pyridine ≥95%
2-Fluoro-6-(1-methylethoxy)pyridine
Precaución: For Research Use Only


Formula: 155.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14599976
CAS Number: 1249358-97-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty