Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one ≥95%

Supplier: AOB CHEM USA
Warning

233212-5G 233212-25G 233212-1G 233212-500MG
77634-432EA 295 USD InStock
77634-432 77654-280 77641-326 77645-308
1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one ≥95%
1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one
Precaución: For Research Use Only


Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD01718113
CAS Number: 14046-55-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty