Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-6-fluoronicotinic acid ≥95%

Supplier: AOB CHEM USA
Warning

24182-5G 24182-1G 24182-25G 24182-500MG
77634-458EA 371 USD InStock
77634-458 77637-404 77658-160 77645-392
5-Chloro-6-fluoronicotinic acid ≥95%
5-Chloro-6-fluoronicotinic acid
Precaución: For Research Use Only


Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426923
CAS Number: 38185-57-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty