Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-chloro-2,6-difluorophenylboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

25597-5G 25597-1G 25597-25G 25597-500MG
77632-278EA 841 USD InStock
77632-278 77637-498 77658-244 77649-548
3-Bromo-5-chloro-2,6-difluorophenylboronic acid ≥95%
3-Bromo-5-chloro-2,6-difluorophenylboronic acid
Precaución: For Research Use Only


Formula: 271.252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24848708
CAS Number: 1451393-03-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty