Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydro-3-(3-methoxyphenyl)-3-furanol ≥95%

Supplier: AOB CHEM USA
Warning

268492-5G 268492-25G 268492-1G 268492-500MG
77628-322EA 1726.28 USD InStock
77628-322 77635-576 77641-634 77645-588
Tetrahydro-3-(3-methoxyphenyl)-3-furanol ≥95%
Tetrahydro-3-(3-methoxyphenyl)-3-furanol
Precaución: For Research Use Only


Formula: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18332787
CAS Number: 1339456-75-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty