Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trimethylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

300121-5G 300121-1G 300121-25G 300121-500MG
77634-676EA 439 USD InStock
77634-676 77637-886 77654-822 77649-900
3,4,5-Trimethylbenzoic acid ≥97%
3,4,5-Trimethylbenzoic acid


Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00184980
CAS Number: 1076-88-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty