Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,6-dimethoxypyridin-4-ylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

30730-5G 30730-500MG 30730-25G 30730-1G
77632-596EA 871 USD InStock
77632-596 77652-622 77658-592 77641-886
3-Chloro-2,6-dimethoxypyridin-4-ylboronic acid ≥97%
3-Chloro-2,6-dimethoxypyridin-4-ylboronic acid
Precaución: For Research Use Only


Formula: 217.415 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088181
CAS Number: 2121512-94-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty