Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Phenoxynicotinic acid methyl ester ≥95%

Supplier: AOB CHEM USA
Warning

32262-1G 32262-500MG 32262-5G
77638-060EA 514 USD InStock
77638-060 77652-646 77658-720
5-Phenoxynicotinic acid methyl ester ≥95%
5-Phenoxynicotinic acid methyl ester
Precaución: For Research Use Only


Formula: 229.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20038719
CAS Number: 936344-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty