Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,4-dimethoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

32344-5G 32344-25G 32344-1G 32344-500MG
77632-716EA 721 USD InStock
77632-716 77655-022 77642-046 77646-056
5-Chloro-2,4-dimethoxyphenylboronic acid pinacol ester ≥97%
5-Chloro-2,4-dimethoxyphenylboronic acid pinacol ester
Precaución: For Research Use Only


Formula: 298.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529808
CAS Number: 2121512-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty