Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

41964-5G 41964-1G 41964-25G 41964-500MG
77635-004EA 334 USD InStock
77635-004 77638-628 77659-210 77650-634
2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester ≥97%
2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester


Formula: C₁₂H₁₂BF₅O₂
Storage Temperature: Ambient
MDL Number: MFCD12405352
CAS Number: 325142-81-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty