Spencer Stitch Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
500380 500380-G 501703 501703-G 501704 501704-G
77662-336EA 46.22 USD InStock
77662-336 77662-338 77662-664 77662-666 77662-668 77662-670
Spencer Stitch Scissors
Scissors
Spencer Stitch Scissors are stitch-cutting scissors or suture scissors for removing sutures or stitches. Spencer Stitch Scissors have a relatively short, straight, and narrow blades.
  • Stich removal scissors
  • 9.0 to 13.25 cm long
  • Straight, blunt tips
  • Notched blade for a sure grip of the suture to cut
  • Surgical grade stainless steel or German steel
The tips of Spencer stitch scissors are blunt rather than sharp to minimize the risk of accidentally puncturing or damaging underlying tissues while cutting sutures. One blade is notched to provide improved grip and traction when cutting through sutures, preventing slippage and ensuring clean cuts of just the suture. The ring handles give you a firm, comfortable grip for precision control.
They come in a variety of lengths, from 9.0 to 13.25 cm. Shorter handles typically offer more control, and longer handles give you greater reach. These Spencer Stitch Scissors are manufactured from surgical grade stainless steel or premium German steel, both of which are autoclavable.
Recommended a properly cleaning of all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty