PCR Plates with Half Skirt - High Profile
Supplier: SARSTEDT INC
72.1979 72.1979.102 72.1979.700
77588-602CS 483.41 USD InStock
77588-602 77588-608 77588-612
PCR Plates with Half Skirt - High Profile
Microplates PCR Plates
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
  • Volume: 0.2 ml
  • 96-well, high profile design
  • Polypropylene material
  • Optimized white color variant for maximum detection sensitivity in qPCR
  • Constant well and plate uniformity minimizes data variability
  • Low DNA-binding variant for highest sample recovery
SARSTEDT PCR plates are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors. For the absolutely reliable absence of DNA and other biomolecules, Biosphere® plus certified products are subjected to a validated decontamination procedure. Biosphere® plus products are also free from DNA, DNase/ RNase, PCR inhibitors, ATP, non-pyrogenic/endotoxin-free and are supplied sterile.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty