SafeSeal Reaction Tubes
Supplier: SARSTEDT INC
72.706 72.706.001 72.706.002 72.706.004 72.706.005 72.706.006 72.706.200 72.706.201 72.706.400
77588-912CS 171.91 USD InStock
77588-912 77588-914 77588-916 77588-918 77588-920 77588-922 77588-924 77588-926 77588-928
SafeSeal Reaction Tubes
Tubes PCR Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • SafeSeal polypropylene caps attached
  • Polypropylene tube material
  • 3 purity grades to meet all requirements
  • Secure seal even under thermal and mechanical stress
  • High centrifugation up to 30,000 ×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base, with or without skirted base, with push cap or screw cap, with or without print or labels.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty