CryoPure Boxes, Flat
Supplier: SARSTEDT INC
93.872.225 93.872.425 93.872.625
77589-732CS 170.47 USD InStock
77589-732 77589-734 77589-736
CryoPure Boxes, Flat
Racks Cryogenic Box Racks
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Suitable for use with 1.2 to 2.0 ml CryoPure tubes, internal and external thread
  • Slip-on lid closure
  • Format: 5×5
  • 25 tube storage capacity
  • Autoclavable (121 °C, 20 minutes)
High-quality polycarbonate storage boxes for storing CryoPure tubes at temperatures down to −196 °C. Numerical codingMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty