PCR Single Tubes - High Profile
Supplier: SARSTEDT INC
72.735.002 72.735.100 72.735.992 72.737 72.737.002 72.737.992
77588-986CS 134.5 USD InStock
77588-986 77588-988 77588-990 77588-992 77588-994 77588-996
PCR Single Tubes - High Profile
Tubes PCR Tubes
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
  • High profile design
  • Flat cap closures attached
  • Polypropylene material
  • Optimal heat transfer through thin-walled reaction chamber
  • Integrated contamination protection; can be opened without touching the inside surface of the lid
SARSTEDT PCR tubes are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors. For the absolutely reliable absence of DNA and other biomolecules, Biosphere® plus certified products are subjected to a validated decontamination procedure. Biosphere® plus products are also free from DNA, DNase/RNase, PCR inhibitors, ATP, non-pyrogenic/endotoxin-free and are supplied sterile. Assorted color option include a mix of Red, Green, Yellow, Blue, Purple, and Orange.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty