Block Racks D13
Supplier: SARSTEDT INC
93.853.132 93.853.133 93.853.134 93.853.135 93.853 93.853.131
77589-702CS 449.85 USD InStock
77589-702 77589-704 77589-706 77589-708 77589-698 77589-700
Block Racks D13
Racks Tube Racks
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Format: 5×10 configuration
  • 13mm opening diameter
  • Without handle
  • Can be autoclaved at 121 °C
  • Range of colors for each area of the laboratory
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.
The Block Rack D13 is designed for space-saving sample storage, and can be combined to form sample distribution blocks. This rack features 50 apertures for S-Monovettes and tubes with 8 to 13 mm diameter.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty