Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-chloro-1-fluoro-4-methoxybenzene ≥95%

Supplier: AOB CHEM USA
Warning

459559-5G 459559-500MG 459559-25G 459559-1G
77633-212EA 912 USD InStock
77633-212 77652-848 77655-862 77642-956
2-Bromo-3-chloro-1-fluoro-4-methoxybenzene ≥95%
2-Bromo-3-chloro-1-fluoro-4-methoxybenzene
Precaución: For Research Use Only


Formula: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD29043796
CAS Number: 1785613-56-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty